ContactUsLinkComponent
Masterflex® Adapter Fittings, Luer to Threaded, Avantor®
Masterflex® Adapter Fittings, Luer to Threaded, Avantor®
Certificates
undefined
Masterflex® Adapter Fittings, Luer to Threaded, Avantor®
Certificates
Specifications
  • Model:
  • 45505-86
  • 45505-88
  • 45502-66
  • 45505-82
  • 45505-84
  • 45505-78
  • 45505-80
  • 45505-76
  • 45502-60
  • Connection type (port 1):
  • Male luer lock
  • Male luer lock
  • Female luer
  • Male luer lock
  • Male luer lock
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • Connection type (port 2):
  • 1/8-27 NPT
  • 1/4" NPT
  • 1/4-28 UNF
  • 1/4-28 UNF
  • 10-32 UNF
  • 1/8-27 NPT
  • 1/8-27 NPT
  • 1/4-28 UNF Bottom Sealing Thread
  • 10-32 UNF
  • No. of ports:
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • Warranty:
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months