ContactUsLinkComponent
Seametrics Low-Flow Impeller Sensors
Seametrics Low-Flow Impeller Sensors
Certificates
undefined
Seametrics Low-Flow Impeller Sensors
Specifications
  • Accuracy:
  • ±1% full-scale
  • ±1% full-scale
  • ±1% full-scale
  • Max. operating temperature:
  • 71 °C (160 °F)
  • 71 °C (160 °F)
  • 71 °C (160 °F)
  • Pressure drop at max. flow:
  • 24 psid
  • 15 psid
  • 20 psid
  • Repeatability:
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • Warranty:
  • 1 year
  • 1 year
  • 1 year