ContactUsLinkComponent
Masterflex® Smart Gasket® 3-Port Sanitary Gaskets, Silicone, Avantor®
Masterflex® Smart Gasket® Sanitary Sampling Gasket, Silicone, 3 Port, 3" Tri-Clamp; 1/EA
Catalog # MFLX30544-20
undefined
Masterflex® Smart Gasket® Sanitary Sampling Gasket, Silicone, 3 Port, 3" Tri-Clamp; 1/EA
Catalog # MFLX30544-20
Specifications
  • Sanitary clamp size (Port 1):
    3"
  • Description:
    Sanitary Sampling Gasket, Silicone, 3 Port, 3" Tri-Clamp; 1/EA
  • Pk:
    1
  • Fitting material:
    Platinum-Cured Silicone
  • Sanitary gasket size:
    3"
  • Warranty:
    1 year
Specifications
About this item
Easily sample fluid in critical high-purity system.
  • Three internal ports accept thermocouple probe0
Masterflex® Smart Gasket® is ideal when validating sterility in a high-purity pharmaceutical system. The Smart Gasket is used to obtain the critical thermal mapping information you need during the validation process. It easily installs between two sanitary standard ferrules.
  • Sanitary clamp size (Port 1):
    3"
  • Description:
    Sanitary Sampling Gasket, Silicone, 3 Port, 3" Tri-Clamp; 1/EA
  • Environmentally Preferable:
  • Pk:
    1
  • Fitting material:
    Platinum-Cured Silicone
  • Sanitary gasket size:
    3"
  • Warranty:
    1 year