ContactUsLinkComponent
Masterflex® Self-Priming Micro Pumps, Avantor®
Masterflex® Self-Priming Micro Pumps, Avantor®
Certificates
undefined
Masterflex® Self-Priming Micro Pumps, Avantor®
Specifications
  • Model:
  • 73120-36
  • 73120-38
  • 73120-32
  • 73120-34
  • 73120-16
  • 73120-30
  • 73120-12
  • 73120-14
  • 73120-10
  • 73120-40
  • 73120-24
  • 73120-22
  • 73120-20
  • 73120-64
  • Inlet connection:
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • Max. temperature:
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • Outlet connection:
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • 1/4"-28 UNF(F)
  • Power pigtail connection:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Run dry:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Self-priming:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Suction lift, wet:
  • 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
  • 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
  • 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
  • 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
  • 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
  • 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
  • 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
  • 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
  • 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
  • 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
  • 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
  • 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
  • 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
  • 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
  • Wetted materials:
  • PTFE, Perfluorelastomer
  • PTFE, Perfluorelastomer
  • PTFE, Perfluorelastomer
  • PTFE, Perfluorelastomer
  • PPS, EPDM
  • PTFE, Perfluorelastomer
  • PPS, EPDM
  • PPS, EPDM
  • PPS, EPDM
  • PTFE, Perfluorelastomer
  • PPS, EPDM
  • PPS, EPDM
  • PPS, EPDM
  • PTFE, Perfluorelastomer
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years