ContactUsLinkComponent
CPC® Twist-to-Connect Quick-Disconnect Fittings, Couplers and Accessories
CPC® Twist-to-Connect Quick-Disconnect Fittings, Couplers and Accessories
Certificates
undefined
CPC® Twist-to-Connect Quick-Disconnect Fittings, Couplers and Accessories
Specifications
  • Max. operating pressure:
  • 35 psi
  • 35 psi
  • 35 psi
  • 35 psi
  • Max. temperature:
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • Min. temperature:
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • Seal material:
  • Platinum-Cured Silicone
  • Platinum-Cured Silicone
  • Platinum-Cured Silicone
  • Sterilisation:
  • Autoclave or gamma
  • Autoclave or gamma
  • Autoclave or gamma
  • Autoclave or gamma
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year