About this item
Superior quality pumps at affordable prices.
- Flow rates from 6 to 56.8 L/hr
- Features double-ball inlet and outlet cartridge-type check valves and heavy-duty, precision molded polypropylene pump head
- Combine engineering excellence with quality materials of construction
- Dual output adjustment, stroke
Digital units offer pinpoint accuracy. They feature an LED readout, 4 to 50 mA, and a 4000:1 turndown ratio. The variable units offer the same digital control with the additional variable speed control capability.
- Environmentally Preferable:
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are: