ContactUsLinkComponent
CPC® APC Acetal Quick-Disconnect Fittings, Bodies and Inserts
undefined
CPC® APC Acetal Quick-Disconnect Fittings, Bodies and Inserts
  • Max. operating pressure:
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • Max. temperature:
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • Min. temperature:
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • Seal material:
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Valve spring material:
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together