ContactUsLinkComponent
Parker Hannifin Panel-Mount Needle Valves, PFA
Parker Hannifin Panel-Mount Needle Valves, PFA
Certificates
undefined
Parker Hannifin Panel-Mount Needle Valves, PFA
Specifications
  • Max. pressure:
  • 2,7 bar
  • 2,7 bar
  • 2,7 bar
  • 2,7 bar
  • Max. temperature:
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • Tubing ext.Ø:
  • 0,5"
  • 0,25"
  • 0,5"
  • 0,375"
  • Wetted materials:
  • PFA
  • PFA
  • PFA
  • PFA
  • Working pressure:
  • 40 psi
  • 40 psi
  • 40 psi
  • 40 psi
  • Warranty:
  • 18 months
  • 1 year
  • 1 year
  • 1 year