ContactUsLinkComponent
CPC® Quick-Disconnect Fittings, 1⁄4-28 Flat-Bottom Port Bodies
CPC® Quick-Disconnect Fittings, 1⁄4-28 Flat-Bottom Port Bodies
Certificates
undefined
CPC® Quick-Disconnect Fittings, 1⁄4-28 Flat-Bottom Port Bodies
Specifications
  • Max. operating pressure:
  • 120 psi
  • 120 psi
  • Max. temperature:
  • 71 °C (160 °F)
  • 71 °C (160 °F)
  • Max. vacuum:
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • Seal material:
  • EPDM
  • EPDM
  • Sterilisation:
  • Gamma
  • Gamma
  • Valve spring material:
  • 316 Stainless Steel
  • 316 Stainless Steel
  • Warranty:
  • 1 year
  • 1 year