ContactUsLinkComponent
CPC® Sanitary Steam-in-Place Connectors, Two-Cycle
undefined
CPC® Sanitary Steam-in-Place Connectors, Two-Cycle
  • Max. operating pressure:
  • 20 psi
  • 20 psi
  • 20 psi
  • Max. temperature:
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • Min. temperature:
  • 4 °C (39 °F)
  • 4 °C (39 °F)
  • 4 °C (39 °F)
  • Seal material:
  • Platinum-cured Silicone
  • Platinum-cured Silicone
  • Platinum-cured Silicone
  • Sterilisation:
  • Gamma or steam-in-place (SIP)
  • Gamma or steam-in-place (SIP)
  • Gamma or steam-in-place (SIP)
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together