ContactUsLinkComponent
Warren Rupp Surge Suppressors and Dampeners
Warren Rupp Surge Suppressors and Dampeners
Certificates
undefined
Warren Rupp Surge Suppressors and Dampeners
Specifications
  • Inlet connection:
  • 1" Tri-Clamp®
  • 1" Tri-Clamp®
  • 0,5 NPT(F)
  • Outlet connection:
  • 1" Tri-Clamp®
  • 1" Tri-Clamp®
  • 0,5 NPT(F)
  • Wetted materials:
  • Aluminum, PTFE
  • Polypropylene, PTFE
  • Polypropylene, PTFE
  • Warranty:
  • 5 years
  • 5 years
  • 5 years