ContactUsLinkComponent
Masterflex® Solenoid-Operated Pinch Valve For Masterflex® Pump Tubing, Avantor®
Masterflex® Solenoid-Operated Pinch Valve For Masterflex® Pump Tubing, Avantor®
Certificates
undefined
Masterflex® Solenoid-Operated Pinch Valve For Masterflex® Pump Tubing, Avantor®
Specifications
  • Power:
  • 12 VDC, 10 W
  • 24 VDC, 10 W
  • 12 VDC, 10 W
  • 12 VDC, 10 W
  • 12 VDC, 10 W
  • 24 VDC, 10 W
  • 12 VDC, 10 W
  • 12 VDC, 10 W
  • 24 VDC, 10 W
  • 12 VDC, 10 W
  • 12 VDC, 10 W
  • 24 VDC, 10 W
  • 24 VDC, 10 W
  • 24 VDC, 10 W
  • 24 VDC, 10 W
  • Response time:
  • 30 - 50 ms
  • 30 - 50 ms
  • 30 - 50 ms
  • 30 - 50 ms
  • 30 - 50 ms
  • 30 - 50 ms
  • 30 - 50 ms
  • 30 - 50 ms
  • 30 - 50 ms
  • 30 - 50 ms
  • 30 - 50 ms
  • 30 - 50 ms
  • 30 - 50 ms
  • 30 - 50 ms
  • 30 - 50 ms
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year