Masterflex® Single-Use Gamma Irradiated Sampling Bottle Assemblies, Sterile, Avantor®
Safely transfer, sample, and store biopharmaceutical products and reagents.
- Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to lose connections
- Assemblies are pre-sterilized by gamma irradiation—eliminates labor and validation costs associated with assembly and cleaning
Cap assembly includes a seamless liner with premolded C-Flex® fill and vent tubes to create an unobstructed fluid path. The 18" fill tube is plugged at the terminal end to maintain sterility until use. Vent tube includes a 0.2-micron PTFE vent filter to prevent infiltration by contaminants. Vent filter is hydrophobic to guard against loss of costly product or media during filling or emptying. The one-piece liner eliminates the risk of bacterial entrapment and contamination and forms an elastomeric seal between cap and bottle rim.
C-Flex tubing is manufactured according to cGMP and meets USP Class VI and USDA requirements. Tubing can withstand temperatures from –100 to 275 °F (–73 to 135 °C). C-Flex tubing has exceptional tensile and tear strength, extremely low extractables, and excellent biocompatibility. The smooth inner surface reduces absorption and protein binding.
: Includes clear PETG bottle, 53B cap, C-Flex® tube/liner assembly with 1/8" ID x 1/4" OD tubing, and 0.2-micron vent filter.
Safely transfer, sample, and store biopharmaceutical products and reagents.
- Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to lose connections
- Assemblies are pre-sterilized by gamma irradiation—eliminates labor and validation costs associated with assembly and cleaning
Cap assembly includes a seamless liner with premolded C-Flex® fill and vent tubes to create an unobstructed fluid path. The 18" fill tube is plugged at the terminal end to maintain sterility until use. Vent tube includes a 0.2-micron PTFE vent filter to prevent infiltration by contaminants. Vent filter is hydrophobic to guard against loss of costly product or media during filling or emptying. The one-piece liner eliminates the risk of bacterial entrapment and contamination and forms an elastomeric seal between cap and bottle rim.
C-Flex tubing is manufactured according to cGMP and meets USP Class VI and USDA requirements. Tubing can withstand temperatures from –100 to 275 °F (–73 to 135 °C). C-Flex tubing has exceptional tensile and tear strength, extremely low extractables, and excellent biocompatibility. The smooth inner surface reduces absorption and protein binding.
: Includes clear PETG bottle, 53B cap, C-Flex® tube/liner assembly with 1/8" ID x 1/4" OD tubing, and 0.2-micron vent filter.
- Special feature:
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile
- Int. Ø tubing:
- 1/8" ID x 1/4" OD
- 1/8" ID x 1/4" OD
- 1/8" ID x 1/4" OD
- 1/8" ID x 1/4" OD
- 1/8" ID x 1/4" OD
- Warranty:
- 3 Months
- 3 Months
- 3 Months
- 3 Months
Product Family Options
- CapacityDescriptionMaterialBottle colourCap colourCap Size
- 125 mlGamma Irradiated Sampling Bottle Assembly, Sterile, PETG Bottle; 125mLPolyethylene terephtalate (PET)ClearNatural53B1 * 1 ST120,00 €CE certificateSterile
Specifications
Cat. No.MFLX95644-00Special featureSterileInt. Ø tubing1/8" ID x 1/4" ODWarranty3 Months - 250 mlGamma Irradiated Sampling Bottle Assembly, Sterile, PETG Bottle; 250mLPolyethylene terephtalate (PET)ClearNatural53B1 * 1 ST141,00 €CE certificateSterile
Specifications
Cat. No.MFLX95644-02Special featureSterileInt. Ø tubing1/8" ID x 1/4" ODWarranty3 Months - 1000 mlGamma Irradiated Sampling Bottle Assembly, Sterile, PETG Bottle; LPolyethylene terephtalate (PET)ClearNatural53B1 * 1 ST161,00 €CE certificateSterile
Specifications
Cat. No.MFLX95644-06Special featureSterileInt. Ø tubing1/8" ID x 1/4" ODWarranty3 Months - 2000 mlGamma Irradiated Sampling Bottle Assembly, Sterile, PETG Bottle; 2LPolyethylene terephtalate (PET)ClearNatural53B1 * 1 ST289,00 €CE certificateSterile
Specifications
Cat. No.MFLX95644-08Special featureSterileInt. Ø tubing1/8" ID x 1/4" ODWarranty - 500 mlGamma Irradiated Sampling Bottle Assembly, Sterile, PETG Bottle; 500mLPolyethylene terephtalate (PET)ClearNatural53B1 * 1 ST126,00 €CE certificateSterile
Specifications
Cat. No.MFLX95644-04Special featureSterileInt. Ø tubing1/8" ID x 1/4" ODWarranty3 Months