ContactUsLinkComponent
Masterflex® I/P® Easy-Load Tubing Retainer Kit, Avantor®
Masterflex® I/P® Replacement Tubing Retainer Kit, PPS/PPS
Catalog # MFLX77602-06
undefined
Masterflex® I/P® Replacement Tubing Retainer Kit, PPS/PPS
Catalog # MFLX77602-06
Specifications
  • Pump series:
    I/P
  • Description:
    I/P® Replacement Tubing Retainer Kit, PPS/PPS
  • Warranty:
    3 Months
  • Pk:
    1
Specifications
About this item
Optimize performance of your I/P® Easy-Load® pump head.
  • Constructed of polyphenylene sulfide (PPS)
Kit includes two polyphenylene sulfide (PPS) retainer assemblies, back plates, and assembly screws.
  • Pump series:
    I/P
  • Description:
    I/P® Replacement Tubing Retainer Kit, PPS/PPS
  • Environmentally Preferable:
  • Warranty:
    3 Months
  • Pk:
    1