ContactUsLinkComponent
Masterflex® Liquid Flow Switches for Threaded Plastic Piping, Avantor®
Masterflex® Liquid Flow Switches for Threaded Plastic Piping, Avantor®
Certificates
undefined
Masterflex® Liquid Flow Switches for Threaded Plastic Piping, Avantor®
Specifications
  • Model:
  • 32774-60
  • 32774-66
  • 32774-64
  • 32774-68
  • 32774-62
  • Max. pressure:
  • 100 psi
  • 100 psi
  • 100 psi
  • 100 psi
  • 100 psi
  • Max. operating temperature:
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • Wetted materials:
  • polypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steel
  • polypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steel
  • polypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steel
  • polypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steel
  • polypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steel
  • Process connection:
  • 0,75 NPT(F)
  • 0,75 NPT(F)
  • 0,75 NPT(F)
  • 0,75 NPT(F)
  • 0,75 NPT(F)
  • Height:
  • 40749"
  • 40749"
  • 40749"
  • 40749"
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year