ContactUsLinkComponent
Masterflex® Inert In-Line Check Valves, PEEK, Avantor®
Masterflex® Inert In-Line Check Valves, PEEK, Avantor®
Certificates
undefined
Masterflex® Inert In-Line Check Valves, PEEK, Avantor®
Specifications
  • Model:
  • 01355-00
  • 01355-04
  • 01355-12
  • 01355-16
  • 01355-24
  • 01355-28
  • Max. pressure:
  • 100 psi
  • 100 psi
  • 100 psi
  • 100 psi
  • 100 psi
  • 100 psi
  • Wetted materials:
  • PEEK/EPDM
  • PEEK
  • PEEK/EPDM
  • PEEK
  • PEEK/EPDM
  • PEEK
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years