Orders LinkContactUsLinkComponent
Multirack bulk tips, Axygen®
Multirack bulk tips, Axygen®
Supplier:   Corning
Multirack bulk tips, Axygen®
Supplier:  Corning
Certificates
Multirack bulk tips, Axygen®
Multirack bulk tips, Axygen®
3 Options Available Below
Product Details & Documents
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an inter-changeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Nonpyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
Axygen® MultiRack bulk pipette tips are compatible with MultiRack racks and packaged in resealable polyethylene bags, non sterile.
Informations de commande: Axygen® MultiRack pipette tips are also available in hinged rack, filter tips, reload, and tipstation configurations.
Product Family Options
Product Information
  • Tip style
    Volume
    Availability
    Price
  • Extended length
    0,1 - 10 µl
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 384,00 €
    384,00 €
    1 * 10.000 ST
       384,00 €
  • Standard length
    1 - 200 µl
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 379,00 €
    379,00 €
    1 * 10.000 ST
       379,00 €
  • Extended length
    50 - 1250 µl
    Log in for availability
    Log in for availability
    1 * 7.680 ST
    1 * 7.680 ST - 391,00 €
    391,00 €
    1 * 7.680 ST
       391,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.