Order Entry
Belgium
ContactUsLinkComponent
10X TAE buffer, liquid, Corning®
10X TAE buffer, liquid, Corning®
Catalog # MDTC46-010-CM
Supplier:  Corning
CAS Number:  
undefined
10X TAE buffer, liquid, Corning®
Catalog # MDTC46-010-CM
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    6
  • Description:
    10X TAE buffer, liquid, 1 L
  • Storage temperature:
    Ambient
  • Shipping temperature:
    Ambient

Specifications

About this item

Tris-Acetate-EDTA (TAE) buffer is used in DNA agarose gel electrophoresis, both in the agarose gel itself and the running buffer

Linear, double-stranded DNA separates faster in a TAE buffer-based system compared to Tris-Borate-EDTAA (TBE) buffer, but the latter has a higher buffering capacity and is thus more resistant to changes in pH. Use of TAE vs. TBE buffer depends on downstream applications. TAE is recommended for preparative electrophoresis in which the nucleic acid bands being separated are to be used in cloning, and other work requiring enzymatic applications.