Orders LinkContactUsLinkComponent
Corning® 25-Layer CellCube® Module, Corning
Corning® 25-Layer CellCube® Module, Corning
   Sustainable Options Available
Supplier:   Corning
Corning® 25-Layer CellCube® Module, Corning
Supplier:  Corning
   Sustainable Options Available
Certificates
Corning® 25-Layer CellCube® Module, Corning
3 Options Available Below
Product Details & Documents
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.
It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth. These CellCube Modules provide: a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates, 21,250 cm² polystyrene growth surface optimally treated for the growth of attachment dependent cells, a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen, an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. Square Flask Shape.
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • CORN3201
  • CORN3304
  • CORN3241
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Media volume:
  • 1500 ml
  • 1500 ml
  • Surface:
  • Tissue Culture Treated
  • Corning® CellBIND®
  • Environmental certification:
  • RECs, ISO 14001 - Kennebunk
Specifications
Product Family Options
Product Information
  • Growth area
    Availability
    Price
  • CORN3201
    25-layer CellCube® module with 21,250 cm² growth surface, Polycarbonate/Polystyrene, Sterile
    25-layer CellCube® module with 21,250 cm² growth surface, Polycarbonate/Polystyrene, Sterile
    21,250 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 698,00 €
    698,00 €
    1 * 1 ST
       698,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CORN3201
    Environmentally Preferable
    Low Manufacturing Impact
    Media volume
    1500 ml
    Surface
    Tissue Culture Treated
    Environmental certification
    RECs, ISO 14001 - Kennebunk
      Low Manufacturing Impact
  • CORN3304
    25-layer CellCube® module with 21,250 cm² growth surface, Polycarbonate/Polystyrene, Sterile
    25-layer CellCube® module with 21,250 cm² growth surface, Polycarbonate/Polystyrene, Sterile
    21,250 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 430,00 €
    1 430,00 €
    1 * 1 ST
       1 430,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    CORN3304
    Environmentally Preferable
    Media volume
    1500 ml
    Surface
    Corning® CellBIND®
    Environmental certification
  • CORN3241
    Corning CellCube®, 25-layer, CellBIND® Surface, Polystyrene with 6"
    Corning CellCube®, 25-layer, CellBIND® Surface, Polystyrene with 6"
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 913,00 €
    913,00 €
    1 * 5 ST
       913,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    CORN3241
    Environmentally Preferable
    Media volume
    Surface
    Environmental certification