Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Supplier: Corning
Certificates
About this item
75 mm Transwell® with 0,4 µm pore polycarbonate membrane insert, sterile.
- 10 µm thick translucent polycarbonate membrane
- Treated for optimal cell attachment
- Each insert is packaged in a 100 mm dish, 12 inserts per case
- Membrane must be stained for cell visibility
Produces a cell culture environment that closely resembles an in vivo state. An essential tool for the study of both anchorage-dependent and independent cell lines.
Specifications
- Catalog No:
- 734-1570
- 734-1569
- 734-1574
- 734-1564
- 734-1563
- 734-1568
- 734-1567
- 734-1573
- CORN7910
- 734-1572
- 734-1576
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk