Order Entry
Belgium
Orders LinkContactUsLinkComponent
10X TBE buffer, liquid, Corning®

10X TBE buffer, liquid, Corning®

10X TBE buffer, liquid, 100 ml, 6
Catalog # MDTC46-011-CM
Supplier:   Corning
10X TBE buffer, liquid, Corning®
Catalog # MDTC46-011-CM
Supplier:  Corning
CAS Number:  
10X TBE buffer, liquid, Corning®
10X TBE buffer, liquid, Corning®

10X TBE buffer, liquid, 100 ml, 6

CAS Number:  
Price
316,00 €
Log in for availability
1 * 6 ST
Product Details & Documents

TBE (Tris-Borate-EDTA) has applications in DNA and RNA agarose gel electrophoresis and polyacrylamide gel electrophoresis.

TBE has a higher buffering capacity than Tris-Acetate-EDTA (TAE) buffer, but the latter is more suitable for steps involving the action of enzymes on nucleic acids because borate is an enzymatic inhibitor. Electrophoresis using TBE-containing agarose gels and TBE running buffer is much more suitable for size analysis of the nucleic acids being separated, as TBE buffer allows these nucleic acids to better maintain their structural integrity.

Specifications

Specifications

  • Pk:
    6
  • Description:
    10X TBE buffer, liquid, 100 ml
  • Cat. No.:
    MDTC46-011-CM
  • Storage temperature:
    Ambient
  • Shipping temperature:
    Ambient

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.