ContactUsLinkComponent
Masterflex® Concentric Reducer Fittings, Sanitary Clamp, Straight, Avantor®
undefined
Masterflex® Concentric Reducer Fittings, Sanitary Clamp, Straight, Avantor®
  • Model:
  • 31805-05
  • 31805-14
  • 31805-10
  • 31805-06
  • Chemical resistance:
  • Excellent
  • Excellent
  • Excellent
  • Very good
  • Excellent
  • Very good
  • Very good
  • Very good
  • Max. operating pressure:
  • 150 psi in ambient conditions, using plastic true union style clamp
  • 150 psi in ambient conditions, using plastic true union style clamp
  • 150 psi in ambient conditions, using plastic true union style clamp
  • 125 psi
  • 150 psi in ambient conditions, using plastic true union style clamp
  • 125 psi
  • 125 psi
  • 125 psi
  • Max. temperature:
  • 138 °C (280 °F)
  • 138 °C (280 °F)
  • 138 °C (280 °F)
  • 82 °C (180 °F)
  • 138 °C (280 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • Min. temperature:
  • -40 °C (-40 °F)
  • -40 °C (-40 °F)
  • -40 °C (-40 °F)
  • –9 °C (15 °F)
  • -40 °C (-40 °F)
  • –9 °C (15 °F)
  • –9 °C (15 °F)
  • –9 °C (15 °F)
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
Frequently Bought Together