ContactUsLinkComponent
Masterflex® Direct-Reading, Variable-Area Flowmeters, Stainless Steel Housing, Avantor®
Masterflex® Direct-Reading, Variable-Area Flowmeters, Stainless Steel Housing, Avantor®
Certificates
undefined
Masterflex® Direct-Reading, Variable-Area Flowmeters, Stainless Steel Housing, Avantor®
Specifications
  • Model:
  • 32447-02
  • 32447-20
  • 32447-00
  • 32447-30
  • 32447-10
  • 32447-54
  • 32447-50
  • 32447-62
  • 32447-60
  • 32447-42
  • 32447-40
  • 32447-32
  • 32447-52
  • 32447-34
  • Accuracy:
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • Float material:
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • Flow Tube material:
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Max. operating temperature:
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • 93 °C (200 °F)
  • O-Ring material:
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Pressure drop at max. flow:
  • 2 water column
  • 6 water column
  • 10 water column
  • 5 water column
  • 24 water column
  • 10 water column
  • 70 water column
  • 39 water column
  • 5 water column
  • 14 water column
  • 13 water column
  • 10 water column
  • Repeatability:
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year