ContactUsLinkComponent
Masterflex® Spring-Loaded Direct-Read Variable-Area Flowmeter, Air/Water/N2, Dual Scale, Avantor®
Masterflex® Spring-Loaded Direct-Read Variable-Area Flowmeter, Air/Water/N2, Dual Scale, Avantor®
Certificates
undefined
Masterflex® Spring-Loaded Direct-Read Variable-Area Flowmeter, Air/Water/N2, Dual Scale, Avantor®
Specifications
  • Model:
  • 32213-26
  • 32211-00
  • 32211-26
  • 32211-12
  • 32211-08
  • 32213-10
  • 32211-18
  • 32211-16
  • 32211-24
  • 32211-22
  • 32211-58
  • 32211-56
  • 32211-60
  • 32211-04
  • 32213-00
  • 32211-02
  • 32211-76
  • Accuracy:
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • Max. operating temperature:
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • 110 °C (230 °F)
  • O-Ring material:
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Pressure drop at max. flow:
  • 4 psid
  • 4 psid
  • 4 psid
  • 4 psid
  • 4 psid
  • 4 psid
  • 4 psid
  • 4 psid
  • 4 psid
  • 4 psid
  • 4 psid
  • 4 psid
  • 4 psid
  • 4 psid
  • 4 psid
  • 4 psid
  • 4 psid
  • Repeatability:
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • ±5% of reading
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year