Order Entry
Belgium
Orders LinkContactUsLinkComponent
Flowline Plastic Mini-Float Level Switches

Flowline Plastic Mini-Float Level Switches

Flowline Plastic Mini-Float Level Switches
Certificates
Flowline Plastic Mini-Float Level Switches

2 Options Available Below

Product Details & Documents

Control liquid level even in corrosive environments.

  • Excellent for clean, non-coating or non-scaling liquids

Choose either vertical or horizontal mounting to fit your application. Reliable reed switch output for interface with PLC or relay control devices. Your liquid must have a specific gravity of 0.8 or greater.

Delivery information: Includes submersible float with 2-ft (0.6-m) cable.

Specifications for Product Family

Specifications

  • Accuracy of depth range:
  • ±5 mm
  • ±5 mm
  • Min. process temperature:
  • -40 °F
  • -40 °F
  • Max. process temperature:
  • 90 °C
  • 90 °C
  • Max. pressure:
  • 25 psi
  • 25 psi
  • Relay output:
  • Reed: 50 VA at 120 VAC/VDC
  • Reed: 50 VA at 120 VAC/VDC
  • Wetted materials:
  • Polypropylene
  • PVDF
  • Warranty:
  • 1 year
  • 1 year

Specifications

Product Family Options

Product Information

  • Power
    Availability
    Price
  • MFLX43299-24
    Float level switches; PP; 1/8" NPT(M)
    Float level switches; PP; 1/8" NPT(M)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,70 €
    63,70 €
    1 * 1 ST
       63,70 €

    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX43299-24
    Accuracy of depth range
    ±5 mm
    Min. process temperature
    -40 °F
    Max. process temperature
    90 °C
    Max. pressure
    25 psi
    Relay output
    Reed: 50 VA at 120 VAC/VDC
    Wetted materials
    Polypropylene
    Warranty
    1 year
    CE Compliant
  • MFLX43299-26
    Mini-Float PVDF Level Switch, SPST, 1/8"NPT(M)
    Mini-Float PVDF Level Switch, SPST, 1/8"NPT(M)
    120 VAC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 130,00 €
    130,00 €
    1 * 1 ST
       130,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    MFLX43299-26
    Accuracy of depth range
    ±5 mm
    Min. process temperature
    -40 °F
    Max. process temperature
    90 °C
    Max. pressure
    25 psi
    Relay output
    Reed: 50 VA at 120 VAC/VDC
    Wetted materials
    PVDF
    Warranty
    1 year
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.