ContactUsLinkComponent
GF Signet Needle Valves with Thread Adapters, PVDF
GF Signet Needle Valves with Thread Adapters, PVDF
Certificates
undefined
GF Signet Needle Valves with Thread Adapters, PVDF
Specifications
  • Max. pressure:
  • 200 bar (200 psi)
  • 200 bar (200 psi)
  • Max. temperature:
  • 150 °C (302 °F)
  • 150 °C (302 °F)
  • Wetted materials:
  • PVDF
  • PVDF
  • Working pressure:
  • 13,8 psi
  • 13,8 psi