ContactUsLinkComponent
Check Valves with Pipe Thread Connections
Check Valves with Pipe Thread Connections
Certificates
undefined
Check Valves with Pipe Thread Connections
Specifications
  • Model:
  • 01350-36
  • 01350-30
  • Max. pressure:
  • 150 psi
  • 100 psi
  • Max. temperature:
  • 60 °C (140 °F)
  • 81 °C (180 °F)
  • Wetted materials:
  • PVDF, Viton
  • Polypropylene, Viton