ContactUsLinkComponent
Nordel Flexible Liner Pumps, Polypropylene
undefined
Nordel Flexible Liner Pumps, Polypropylene
  • Duty cycle continuous:
  • Yes
  • Yes
  • Inlet connection:
  • 1-1/4" hose barb
  • 3/4" hose barb
  • Max. temperature:
  • 85 °C (185 °F)
  • 85 °C (185 °F)
  • Motor hp:
  • 1/2
  • 1/4
  • Outlet connection:
  • 1" hose barb
  • 3/4" hose barb
  • Power conduit box connection:
  • Yes
  • Yes
  • Run dry:
  • Yes
  • Yes
  • Self-priming:
  • Yes
  • Yes
  • Suction lift, dry:
  • 1800 mm of H₂O (72" H₂O); 629 mm Hg (5,3" Hg)
  • 1800 mm of H₂O (72" H₂O); 629 mm Hg (5,3" Hg)
  • Wetted materials:
  • Polypropylene, Nordel®
  • Polypropylene, Nordel®
  • Warranty:
  • 1 year
  • 1 year
Frequently Bought Together