Guzzler® Diaphragm Hand Pump Corregated Hoses
Supplier: Avantor Fluid Handling
Certificates
About this item
Optimize performance of your Guzzler® Diaphragm Hand Pump.
- Suitable for use with Guzzler® Portable Diaphragm Hand Pumps
These hoses are noncollapsing, wire-reinforced, and constructed of durable vinyl.
Model 06309-10 hose is compatible with Guzzler pumps 07090-09 and -10.
Model 06309-11 hose is compatible with Guzzler pump 07090-11.
Model 06309-12 hose is compatible with Guzzler pump 07090-12.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX06309-10
- MFLX06309-11
- Tube length, inlet:
- 488 cm (192")
- 488 cm (192")
- Warranty:
- 1 year
- 1 year