ContactUsLinkComponent
Masterflex® Ismatec® Quick-Couple, Single-Channel Pump Heads for 1.6 or 2.4 mm Wall Tubing, Avantor®
Masterflex® Ismatec® Quick-Couple, Single-Channel Pump Heads for 1.6 or 2.4 mm Wall Tubing, Avantor®
Certificates
undefined
Masterflex® Ismatec® Quick-Couple, Single-Channel Pump Heads for 1.6 or 2.4 mm Wall Tubing, Avantor®
Specifications
  • Model:
  • 78002-22
  • 78002-16
  • 78002-11
  • Max. pressure:
  • 1,5 bar (21,7 psi)
  • 1,5 bar (21,7 psi)
  • 1,5 bar (21,7 psi)
  • Min. flow rate:
  • 0,072 ml/min
  • 0,45 ml/min
  • 0,49 ml/min
  • No. of channels:
  • 1
  • 1
  • 1
  • Series:
  • Quick-Coupled
  • Quick-Coupled
  • Quick-Coupled
  • Warranty:
  • 1 year
  • 2 years
  • 2 years