ContactUsLinkComponent
Masterflex® Replacement RJ-12 Cable for Liqui-Sense Sensor, 6-pin; 3.5 ft
  MFLX77095-02
undefined
Masterflex® Replacement RJ-12 Cable for Liqui-Sense Sensor, 6-pin; 3.5 ft
  MFLX77095-02
  • Description:
    Replacement RJ-12 cable
  • Length:
    3,5 ft.
  • Model:
    77095-02
  • Pk:
    106,68 cm
Make the most of your Liqui-Sense leak detector with suitable replacement parts.
  • Compatible with Masterflex® Liqui-Sense controllers
This replacement cable features a 6-pin cable with RJ-12 connections and has a 3,5 foot cable length.
  • Description:
    Replacement RJ-12 cable
  • Length:
    3,5 ft.
  • Environmentally Preferable:
  • Model:
    77095-02
  • Pk:
    106,68 cm