ContactUsLinkComponent
Masterflex® I/P® Standard Pump Heads, Avantor®
Masterflex® I/P® Standard Pump Heads, Avantor®
Certificates
undefined
Masterflex® I/P® Standard Pump Heads, Avantor®
Specifications
  • Model:
  • 07019-21
  • 07019-31
  • 07019-20
  • 07019-53
  • 07019-43
  • 07019-32
  • Gear ratio:
  • 1:1
  • 1:1
  • 1:1
  • 1:1
  • 1:1
  • 1:1
  • No. of channels:
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • Occlusion:
  • Fixed
  • Fixed
  • Fixed
  • Fixed
  • Fixed
  • Fixed
  • Open-head sensor:
  • No
  • No
  • No
  • No
  • No
  • No
  • Optimum RPM range:
  • 0,1 - 650 rpm
  • 0,1 - 650 rpm
  • 0,1 - 650 rpm
  • 0,1 - 650 rpm
  • 0,1 - 650 rpm
  • 0,1 - 650 rpm
  • Pump series:
  • I/P
  • I/P
  • I/P
  • I/P
  • I/P
  • I/P
  • Tubing retention:
  • Manual
  • Manual
  • Manual
  • Manual
  • Manual
  • Manual
  • Tubing sizes accepted:
  • I/P® 73
  • I/P® 82
  • I/P® 73
  • I/P® 82
  • I/P® 73
  • I/P® 82
  • Volume per revolution with I/P 73 tubing:
  • 12,3 ml
  • 12,3 ml
  • 12,3 ml
  • Volume per revolution with I/P 82 tubing:
  • 20 ml
  • 20 ml
  • 20 ml
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years