ContactUsLinkComponent
Masterflex® Correlated Variable-Area Flowmeters with Valves, High-Flow, 65-mm, Avantor®
Masterflex® Correlated Variable-Area Flowmeters with Valves, High-Flow, 65-mm, Avantor®
Certificates
undefined
Masterflex® Correlated Variable-Area Flowmeters with Valves, High-Flow, 65-mm, Avantor®
Specifications
  • Model:
  • 03216-88
  • 03218-41
  • 03293-40
  • 03218-39
  • 03216-36
  • 03216-90
  • 03293-38
  • 03216-38
  • 03218-37
  • 03216-40
  • Accuracy:
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • Float material:
  • Carboloy
  • Carboloy
  • 316 SS
  • Glass
  • Sapphire
  • 316 SS
  • 316 SS
  • Glass
  • Carboloy
  • Flow Tube material:
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Max. operating temperature:
  • 65 °C (150 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 65 °C (150 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • O-Ring material:
  • PTFE
  • Viton®
  • Buna-N
  • Viton®
  • Buna-N
  • PTFE
  • Buna-N
  • Buna-N
  • Viton®
  • Buna-N
  • Repeatability:
  • ±0,25% full-scale
  • ±0,25%
  • ±0,25%
  • ±0,25%
  • ±0,25%
  • ±0,25%
  • ±0,25%
  • ±0,25%
  • ±0,25%
  • ±0,25%
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year