ContactUsLinkComponent
Masterflex® Direct-Read Variable Area Flowmeters for Air, Graph-Style, Aluminum, Avantor®
Masterflex® Direct-Read Variable Area Flowmeters for Air, Graph-Style, Aluminum, Avantor®
Certificates
undefined
Masterflex® Direct-Read Variable Area Flowmeters for Air, Graph-Style, Aluminum, Avantor®
Specifications
  • Accuracy:
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • Max. operating temperature:
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • Max. pressure:
  • 1000 psi
  • 1000 psi
  • 1000 psi
  • 1000 psi
  • 1000 psi
  • 1000 psi
  • 1000 psi
  • 1000 psi
  • 1000 psi
  • O-Ring material:
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Repeatability:
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year