ContactUsLinkComponent
Masterflex® Reducer Fittings, Hose Barb, Straight, Stainless Steel
Masterflex® Reducer Fittings, Hose Barb, Straight, Stainless Steel
Certificates
undefined
Masterflex® Reducer Fittings, Hose Barb, Straight, Stainless Steel
Certificates
Specifications
  • Model:
  • 30620-11
  • 30620-12
  • 30620-09
  • 31208-14
  • 30620-10
  • 31208-15
  • 31208-13
  • Orifice dimensions:
  • 4,763 mm (0,1875")
  • 8,73 mm (0,34375")
  • 0,7925 mm (0,03125")
  • 4,763 mm (0,1875")
  • 0,7925 mm (0,03125")
  • Sterilisation:
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Warranty:
  • 1 Month
  • 1 Month
  • 1 year
  • 1 Month
  • 1 year
  • 1 year