Orders LinkContactUsLinkComponent
Masterflex® B/T® Drive Controller Mounting Bracket, Avantor®
Masterflex® B/T® Drive Controller Mounting Bracket, Avantor®
Controller mounting bracket for B/T Modular Digital Pump System. 2 years warranty
Catalog # MFLX77111-90
Masterflex® B/T® Drive Controller Mounting Bracket, Avantor®
Catalog # MFLX77111-90
Masterflex® B/T® Drive Controller Mounting Bracket, Avantor®
Masterflex® B/T® Drive Controller Mounting Bracket, Avantor®
Controller mounting bracket for B/T Modular Digital Pump System. 2 years warranty
Price
493,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Make controlling your B/T® drive easier than ever.
  • Suitable for use with B/T modular digital drives
This controller mounting bracket allows a Masterflex® B/T® digital controller to be mounted directly to the pump head for ease of use.
Specifications
  • Description:
    Controller mounting bracket for B/T Modular Digital Pump System. 2 years warranty
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.