ContactUsLinkComponent
Masterflex® Solenoid Valve Connection Cable, Avantor®
Masterflex® Connection cable, "D" connector to stripped ends, 3-ft
Catalog # MFLX98651-50
undefined
Masterflex® Connection cable, "D" connector to stripped ends, 3-ft
Catalog # MFLX98651-50
Specifications
  • Description:
    Connection cable, "D" connector to stripped ends
  • Length:
    3 ft
  • Model:
    98651-50
  • Warranty:
    1 year
  • Pk:
    3 Feet
Specifications
About this item
Easy connect to your solenoid valve.
  • Optimize performance of your Masterflex® Solenoid Valves
"D" connector connection cable with stripped wire ends for Aalborg stepping motor proportioning valves. This cable is 3-ft (0.9m) long.
  • Description:
    Connection cable, "D" connector to stripped ends
  • Length:
    3 ft
  • Environmentally Preferable:
  • Model:
    98651-50
  • Warranty:
    1 year
  • Pk:
    3 Feet