ContactUsLinkComponent
1536-well spheroid microplates, Corning®
Catalog # CORN4637
Supplier:  Corning
   Low Manufacturing Impact
undefined
1536-well spheroid microplates, Corning®
Catalog # CORN4637
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • No. of wells:
    1536
  • Packed:
    1 / Pk, 5 / Cs
  • Recommended working volume:
    5 - 10 µl
  • Sterile:
    +
  • Colour:
    Black
  • Description:
    1536-well black/clear round bottom ultra-low attachment spheroid microplate, with lid, sterile
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Pk:
    5
  • Well volume:
    14 ml
Specifications
About this item
With their proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment (ULA) surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.
  • 536-well format allows for high throughput 3D cell culture
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
  • Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization
  • Unique design shields each well to minimize well-to-well cross-talk
You can culture and assay spheroids in the same microplate, without the need for transfer to a new microplate. Compatible with the GNF ultra high throughput system.