Orders LinkContactUsLinkComponent
Single-use bags for HYPERStack® vessels
Single-use bags for HYPERStack® vessels
Supplier:   Corning
Single-use bags for HYPERStack® vessels
Supplier:  Corning
Certificates
Single-use bags for HYPERStack® vessels
3 Options Available Below
Product Details & Documents
These single-use bags can be connected to tubing via tube welding or by using the pre-assembled multipurpose connectors.
They can be used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution.
Product Family Options
Product Information