Order Entry
Belgium
ContactUsLinkComponent
Serum-free/low-protein medium, 1X with L-glutamine and 1 g/L BSA, without phenol red, Corning®
Serum-free/low-protein medium, 1X with L-glutamine and 1 g/L BSA, without phenol red, Corning®
Catalog # MDTC40-102-CV
Supplier:  Corning
CAS Number:  
undefined
Serum-free/low-protein medium, 1X with L-glutamine and 1 g/L BSA, without phenol red, Corning®
Catalog # MDTC40-102-CV
Supplier:  Corning
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Pk:
    6
  • Description:
    Corning® cellgro® COMPLETE Serum-Free/Low-Protein Medium, 1X with L-Glutamine and 1 g/L BSA without Phenol Red
  • Size:
    500 ml
  • Storage temperature:
    Refrigerate

Specifications

About this item

Corning® cellgro® COMPLETE formulation is based on a combination of 1:1 DMEM/F12, RPMI, and McCoy's 5A medium. This medium contains a proprietary mixture of selected trace elements and high molecular weight carbohydrates, extra vitamins, animal origin-free protein, and a small amount of high quality bovine serum albumin (1,0g/L). The Corning cellgro COMPLETE supports superior growth and viability over long-term passages in hybridomas, along with suspensions and adherent cell cultures.