VWR®, Septa 18 mm
Supplier: VWR Collection
Certificates
About this item
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
- Fits for a broad range of caps
- Easy exchange
- Different hardness and septa thickness available
Septa choice depends on the application.
Frequently Bought Together