Orders LinkContactUsLinkComponent
VWR®, Septa ND11
VWR®, Septa ND11
VWR®, Septa ND11
Supplier:  VWR Collection
Certificates
VWR®, Septa ND11
VWR®, Septa ND11
12 Options Available Below
Product Details & Documents
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available
Septa choice depends on the application.
Product Family Options
Product Information
  • Septum
    Hardness
    Thickness
    Availability
    Price
  • White silicone/red PTFE
    45° shore A
    1,3 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 322,00 €
    322,00 €
    1 * 1.000 ST
       322,00 €
  • Red PTFE/white silicone/red PTFE
    45° shore A
    1,0 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 394,00 €
    394,00 €
    1 * 1.000 ST
       394,00 €
  • Cream silicon/red PTFE
    55° shore A
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 279,00 €
    279,00 €
    1 * 1.000 ST
       279,00 €
  • PTFE virginal
    53° shore D
    0,25 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 91,40 €
    91,40 €
    1 * 1.000 ST
       91,40 €
  • Viton 1A black
    70° shore A
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 702,00 €
    702,00 €
    1 * 1.000 ST
       702,00 €
  • White silicone/blue PTFE, cross-slit
    55° shore A
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 382,00 €
    382,00 €
    1 * 1.000 ST
       382,00 €
  • Red-orange natural rubber/transparent TEF
    60° shore A
    1,0 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 127,00 €
    127,00 €
    1 * 1.000 ST
       127,00 €
  • Grey PTFE/cream butyl/grey PTFE
    55° shore A
    1,3 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 163,00 €
    163,00 €
    1 * 1.000 ST
       163,00 €
  • White silicone/silver aluminium
    50° shore A
    1,3 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 637,00 €
    637,00 €
    1 * 1.000 ST
       637,00 €
  • Red rubber/beige PTFE
    45° shore A
    1,0 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 215,00 €
    215,00 €
    1 * 1.000 ST
       215,00 €
  • Universal HT injection port, transparent
    3,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 40,80 €
    40,80 €
    1 * 25 ST
       40,80 €
  • High performance GC injection port, red, with centre guide
    3,0 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 131,00 €
    131,00 €
    1 * 50 ST
       131,00 €