Accessories for rotary sample changer 7200 - for piston burettes and automatic titrators
SCOI285201994
- Description:Glass titration beaker
- Environmentally Preferable:
- Pk:10
- Details:250 ml tall form (10 pieces) for TZ 1459/TZ 4016/TZ4025 and TZ 3942