Blasticidin S hydrochloride, powder, Corning®
Catalog # MDTC30-100-RB
Supplier: Corning
CAS Number:
Specifications
- Pk:1
- Seq:2370
- Environmentally Preferable:
- Pk Info:50 mg
Specifications
About this item
:
:458.904 g/mol
Storage Temperature:Freezer
CAS Number:3513-03-9
Formula:C₁₇H₂₇ClN₈O₅
MDL Number:MFCD02091640