Fetal bovine serum characterized (United States), HyClone™
Supplier: HyClone products (Cytiva)
Certificates
About this item
Enhance growth of cells with U.S.-sourced Cytiva HyClone™ characterised fetal bovine serum (FBS). More economical than defined FBS, but meets the requirements of most discriminating cell culturists.
Tested for endotoxin, hemoglobin, and additional biochemical assays and filtered through serial 100 nm pore-size rated filters.
Specifications
- Catalog No:
- SH30071.03IH25
- SH30071.03IH30
- Origin:
- US
- US
- Endotoxin:
- ≤25 EU/ml
- ≤25 EU/ml
- Haemoglobin:
- <25 mg/dl
- <25 mg/dl
- Storage temperature:
- <–10 °C
- <–10 °C
Specifications
Frequently Bought Together