Order Entry
Belgium
ContactUsLinkComponent
Hybridoma growth and expression media HYGM6
Hybridoma growth and expression media HYGM6
Catalog # BCON9-00F55-I
Supplier:  BIOCONCEPT
CAS Number:  
undefined
Hybridoma growth and expression media HYGM6
Catalog # BCON9-00F55-I
Supplier:  BIOCONCEPT
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    Hybridoma growth and expression media HYGM6 - liquid
  • Size:
    500 ml bottle
  • Sterility:
    Sterile
  • Shipping conditions:
    Ambient
  • Storage temperature:
    2…4 °C
  • Shelf life:
    9 Months

Specifications

About this item

Hybridoma growth media (HYGM) 6 is a serum-free and fully defined media that can be used for the cultivation of various different hybridomas and the production of monoclonal antibodies.

  • Serum-free
  • Ready to use medium
  • High yielding
  • contsins therapeutic grade recombinant insulin
  • Free of hydrolysates or peptones
  • Free of any undefined components

HYGM-6 medium contains recombinant therapeutic grade insulin. It is the only protein in that medium, no other proteins or undefined hydrolysates are present.

Certifications: ISO 9001; cGMP.