Orders LinkContactUsLinkComponent
Label printer, M210
Label printer, M210
Supplier:   Brady
Label printer, M210
Supplier:  Brady
Certificates
Label printer, M210
2 Options Available Below
Product Details & Documents
The M210 Portable Label Printer is an all-in-one labeller for voice/datacom, electrical and general industrial teams. With label materials designed specifically for harsh environments, the M210 printer can quickly and easily create clear labels that stay stuck for years - despite extreme temperatures and curved or highly textured surfaces.
  • Monocolour
  • Engineered long lasting material quality
  • Prints durable true-sized 6 to 19 mm wide identification to maximise amount of data
  • Adjust the length of the labels using the high performance continuous material
  • Ergonomic cutter and label grabber, holding the label after cutting
  • Easy to use: ABC keypad, graphics library, hot keys and menu functions for fast label creation
  • Smart Cell Technology for automatic label setup and automatic formatting
  • 2 year warranty
Languages available include Croatian; Czech; Danish; Dutch; English(US); Estonian; Finnish; French; German; Hungarian; Italian; Norwegian(Bokmål); Polish; Portuguese; Romanian; Slovak; Slovenian; Spanish (universal); Swedish; Turkish.
Accessories information: Optional multifunctional accessory consists of magnet, flashlight, stand, belt loop and pole strap. 2 AAA batteries required (not included).
Delivery information: The 'carton pack' model ships in a carton pack with a quick start guide and an M21-750-499 label cartridge.
The kits ship in a hard case holding a M210 printer, a Li-ion battery, a EU or UK power cord, and a M21-750-499 label cartridge.
Specifications for Product Family
Specifications
  • Catalog No:
  • BRDY152260
  • BRDY311307
  • Label:
  • Continuous and die-cut
  • Continuous and die-cut
  • Print resolution:
  • 203 dpi
  • 203 dpi
Specifications
Product Family Options
Product Information
  • Print width
    W×D×H
    Availability
    Price
  • BRDY152260
    M210 Label printer, carton pack, no plug
    M210 Label printer, carton pack, no plug
    16,00 mm
    114,30×63,50×241,30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 99,00 €
    99,00 €
    1 * 1 ST
       99,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    BRDY152260
    Label
    Continuous and die-cut
    Print resolution
    203 dpi
  • BRDY311307
    M210 Label printer kit, EU plug
    M210 Label printer kit, EU plug
    16,00 mm
    114,30×63,50×241,30 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 190,00 €
    190,00 €
    1 * 1 KIT
       190,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    BRDY311307
    Label
    Continuous and die-cut
    Print resolution
    203 dpi
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Multifunctional accessory for M210 and M210-LAB label printers
    Multifunctional accessory for M210 and M210-LAB label printers
    BRDY170424
    Log in for availability
    1 * 1 ST - 35,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Hard case for M210 and M210-LAB label printers
    Hard case for M210 and M210-LAB label printers
    BRDY170421
    Log in for availability
    1 * 1 ST - 44,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Belt clip for M210 and M211 label printers
    Belt clip for M210 and M211 label printers
    BRDY170385
    Log in for availability
    1 * 1 ST - 11,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Magnet only accessory for M210 and M210-LAB label printers
    Magnet only accessory for M210 and M210-LAB label printers
    BRDY170425
    Log in for availability
    1 * 1 ST - 16,70 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Brady workstation scan and print software suite
    Brady workstation scan and print software suite
    BRDY149445
    BBP11, BBP12, BBP31, BBP33, BBP35, BBP37, BBP81, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Colour and Cut, GlobalMark 2 Multicolour, i3300, i5100, i7100, i7500, IDXPERT, IP Series, J2000, J4000, J5000, LABXPERT, M410, M510, M610, M611, M710, PR Plus, S3100, THT 3" Core Printers, TLS 2200
    Log in for availability
    1 * 1 ST - 833,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Workstation lockout writer software app
    Workstation lockout writer software app
    BRDY145445
    BBP®31, BBP®33, BBP®85, i3300
    Log in for availability
    1 * 1 ST - 833,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Brady workstation automation software suite
    Brady workstation automation software suite
    BRDY149187
    BBP11, BBP12, BBP30, BBP33, BBP35, BBP37, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Multicolor, i3300, i5100, i7100, IP Series, J2000, J4000, J5000, M610, M611, M710, PR Plus, S3000, THT 76 mm Core Printers, Wraptor, Wraptor A6500
    Log in for availability
    1 * 1 ST - 833,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Brady workstation CLP/GHS app via download
    Brady workstation CLP/GHS app via download
    BRDY195526
    Log in for availability
    1 * 1 ST - 369,99 €