ContactUsLinkComponent
Service and retail wiping paper, centrefeed roll, WypAll® Reach™
Service and retail wiping paper, centrefeed roll, WypAll® Reach™
Catalog # 115-2527
Supplier:  KIMBERLY CLARK
undefined
Service and retail wiping paper, centrefeed roll, WypAll® Reach™
Catalog # 115-2527
Supplier:  KIMBERLY CLARK
Specifications
  • Colour:
    Blue
  • Description:
    Service and retail wiping paper
  • L×W:
    380×183 mm
  • Pk:
    6 Roll
Specifications
About this item
WypAll® L10 centrefeed roll is blue roll designed for light-duty hygiene wiping and designed for use in retail and service environments. Compatible with the portable WypAll® Reach™ dispenser (115-2528) which ensures rolls remain enclosed to help prevent against transmission of food-borne bacteria.
  • Each WypAll® L10 centrefeed blue roll contains 280 embossed wiping paper sheets
  • Designed specifically for the WypAll® Reach™ wiping paper dispenser, using the black dispensing nozzle
  • Wiping Rolls remain clean and enclosed to help prevent against transmission of food-borne bacteria
  • Centrefeed roll format allows for easy single sheet dispensing
  • Exceptional strength and absorbency thanks to the Airflex™ Technology in these disposable wipers
  • Single-use blue roll is designed to be used once with detergent or disinfectant
No matter what surfaces they may come into contact with throughout the day, provide employees and visitors greater peace of mind by offering proven cleaning and disinfecting workplace hygiene solutions to help break the chain of germ transmission.WypAll® L10 professional wiping paper products are designed for the specific environments where they will be used. These centrefeed blue roll disposable paper wipes are 1-ply and are designed for retail and service environments.
WypAll® brand embossing increases sheet strength owing to unique pattern ply-bonds across the entire surface of the sheet, also delivering an enhanced appearance and feel to every sheet of WypAll® L10 blue roll. Made specifically for use with the portable WypAll® Reach™ paper wipes dispenser (115-2528), this easy to handle, portable and enclosed system is designed to keep the centrefeed roll clean, which helps to prevent against transmission of food-borne bacteria. The single sheet dispensing is designed to minimise contamination while encouraging efficient use of the blue roll. The sheet by sheet dispensing reduces paper consumption by over 20% and cuts paper waste by over 20%. The unique Airflex™ Technology in this wiping paper is designed for exceptional strength and absorbency.
These WypAll® paper wipers are designed to be used once with detergent or disinfectant. They have been tested as compatible for use in spray and wipe application with peracetic acid, hydrogen peroxide, and quats (quaternary ammonium compounds). Spray and wiping compatibility helps to ensure workers can wipe work spaces, vehicles, desks, seating, and food prep areas before and after use. Spray and wipe also helps to enable production workers to clean down/sanitise individual workstations before hand-off to the next shift. For environments where fully saturated wiping is a requirement, these WypAll® wiping rolls are tested as compatible with soap/detergent. Always follow manufacturers instructions for dilution, application, and contact time.