Order Entry
Belgium
ContactUsLinkComponent
Dichloromethane ≥99.9% (GC, on anhydrous basis) stabilised for peptide synthesis
Irritant
Health
Dichloromethane ≥99.9% (GC, on anhydrous basis) stabilised for peptide synthesis
Catalog # 89048.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Health
Dichloromethane ≥99.9% (GC, on anhydrous basis) stabilised for peptide synthesis
Catalog # 89048.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (GC on anhydrous basis):
    min. 99.9%
  • Clear colorless liquid:
    complying
  • Color (APHA Pt-Co):
    max. 10
  • Free amines (Kaiser):
    max. 0.001%
  • Residue after evaporation:
    max. 0.0005% (w/w)
  • Water (Karl Fischer):
    max. 0.005% (w/w)
  • Iron (Fe):
    max. 0.1 ppm
  • Magnesium (Mg):
    max. 0.1 ppm
  • Lead (Pb):
    max. 0.1 ppm
  • Zinc (Zn):
    max. 0.1 ppm
  • Stabilizer (Amylene):
    0.003-0.006% (w/w)

Specification Test Results

About this item

 :
 :84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
 :Stabilised with amylene 0.003-0.006 %
REACH:01-2119480404-41
 :GHS07
 :GHS08
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881