Stand-up/sit down tables/workbench, height-adjustable
About this item
The height adjustable stainless steel table is manufactured from fully welded 304-grade stainless steel in a brushed dull polish finish and is available in electronic or manual wind height adjustment.
- Free standing
- 304-grade stainless steel
- Brushed dull polish finish
- Fully welded, no construction or assembly required
- Hydraulic height adjustable table - manual or electric
- Stainless steel feet with non-marking rubber base
- 700 to 1000 mm height adjustment (table top height)
The 300 mm height adjustment, ensures a work surface comfortable for workers of different heights.
Load capacity: 250 kg
6 Options Available Below
Product Details & Documents
The height adjustable stainless steel table is manufactured from fully welded 304-grade stainless steel in a brushed dull polish finish and is available in electronic or manual wind height adjustment.
- Free standing
- 304-grade stainless steel
- Brushed dull polish finish
- Fully welded, no construction or assembly required
- Hydraulic height adjustable table - manual or electric
- Stainless steel feet with non-marking rubber base
- 700 to 1000 mm height adjustment (table top height)
The 300 mm height adjustment, ensures a work surface comfortable for workers of different heights.
Load capacity: 250 kg
Product Family Options
Manual height adjustment
Product Information
- W×D×HAvailabilityPrice
- TEKMETSS1300X750Height adjustable table, no shelf, rear rail
- TEKMTLWB-AJMA16X75Height adjustable table, no shelf, rear rail
- TEKMETSS1900X750Height adjustable table, no shelf, rear rail
Electronic height adjustment
Product Information
- W×D×HAvailabilityPrice
- TEKMETSS1300X750/EHeight adjustable table. electric, no shelf, rear railHeight adjustable table. electric, no shelf, rear rail1300×750×840 mmLog in for availability1 * 1 ST
- TEKMETSS1600X750/EHeight adjustable table, electric, no shelf, rear railHeight adjustable table, electric, no shelf, rear rail1600×750×840 mmLog in for availability1 * 1 ST
- TEKMETSS1900X750/EHeight adjustable table, electric, no shelf, rear railHeight adjustable table, electric, no shelf, rear rail1900×750×840 mmLog in for availability1 * 1 ST